missing translation for 'onlineSavingsMsg'
Få mere at vide

Tocris Bioscience™ GC 1
GREENER_CHOICE

Artikelnummer. 30001660
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30001660 50 mg
30001642 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001660 Supplier Tocris Bioscience™ Supplier No. 4554/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High affinity thyroid receptor α (TRα) and TRβ agonist; thyromimetic

Thyromimetic; high affinity agonist at thyroid hormone receptor (TR) β and TRα receptors (KD values are 67 and 440 pM respectively). Displays 5- and 100-fold greater potency than the endogenous agonist T3 in vitro at TRα1 and TRβ1 receptors respectively. Promotes reverse cholesterol transport in chow- and fat-fed mice. Induces proliferation of hepatocytes and pancreatic acinar cells in rats independent of prior tissue injury.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 211110-63-3
Molekylær formel C20H24O4

Specifications

Kemisk navn eller materiale GC 1
Mængde 50 mg
Formel vægt 328.4
Renhed >98%
Molekylvægt (g/mol) 328.408
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.