missing translation for 'onlineSavingsMsg'
Learn More

GBR 12935 dihydrochloride, Thermo Scientific™

Catalog Number p-7042568
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
67469-81-2
C28H36Cl2N2O
487.509
MFCD00083175
NQWRSILGEXNJIT-UHFFFAOYSA-N
gbr 12935 dihydrochloride, gbr-12935 dihydrochloride, 1-2-diphenylmethoxy ethyl-4-3-phenylpropyl piperazine dihydrochloride, 1-2-diphenylmethoxyethyl-4-3-phenylpropyl piperazine dihydrochloride, 1-2-benzhydryloxyethyl-4-3-phenylpropyl piperazine;dihydrochloride, 1-2-benzhydryloxy ethyl-4-3-phenylpropyl piperazine dihydrochloride, 1-2-diphenylmethoxy ethyl-4-3-phenylpropyl-piperazine dihydrochloride, 1-2-diphenylmethoxy ethyl-4-3-phenylpropyl piperazinediium dichloride, gbr12935 dihydrochloride
11957553
CHEBI:64091
1-(2-benzhydryloxyethyl)-4-(3-phenylpropyl)piperazine;dihydrochloride
C1CN(CCN1CCCC2=CC=CC=C2)CCOC(C3=CC=CC=C3)C4=CC=CC=C4.Cl.Cl
This item is not returnable. View return policy
67469-81-2
C28H36Cl2N2O
487.509
MFCD00083175
NQWRSILGEXNJIT-UHFFFAOYSA-N
gbr 12935 dihydrochloride, gbr-12935 dihydrochloride, 1-2-diphenylmethoxy ethyl-4-3-phenylpropyl piperazine dihydrochloride, 1-2-diphenylmethoxyethyl-4-3-phenylpropyl piperazine dihydrochloride, 1-2-benzhydryloxyethyl-4-3-phenylpropyl piperazine;dihydrochloride, 1-2-benzhydryloxy ethyl-4-3-phenylpropyl piperazine dihydrochloride, 1-2-diphenylmethoxy ethyl-4-3-phenylpropyl-piperazine dihydrochloride, 1-2-diphenylmethoxy ethyl-4-3-phenylpropyl piperazinediium dichloride, gbr12935 dihydrochloride
11957553
CHEBI:64091
1-(2-benzhydryloxyethyl)-4-(3-phenylpropyl)piperazine;dihydrochloride
C1CN(CCN1CCCC2=CC=CC=C2)CCOC(C3=CC=CC=C3)C4=CC=CC=C4.Cl.Cl

A very potent and selective inhibitor of dopamine uptake

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color White
Quantity 10 mg
Formula Weight 487.51
Physical Form Powder
Chemical Name or Material GBR 12935 dihydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'