missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GBR 12909 dihydrochloride, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 mg
50 mg
Description
Dopamine uptake inhibitor
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | White |
Quantity | 10 mg |
Synonym | vanoxerine dihydrochloride, gbr 12909 dihydrochloride, vanoxeamine, gbr-12909 dihydrochloride, vanoxerine hydrochloride, unii-mwo1ip03ev, mwo1ip03ev, vanoxerine dihydrochloride gbr12909, gbr12909 dihydrochloride, 1-2-bis 4-fluorophenyl methoxy ethyl-4-3-phenylpropyl piperazine dihydrochloride |
Solubility Information | Soluble in DMSO to 50mM; Soluble to 5mM in water with warming. |
Formula Weight | 523.49 |
Physical Form | Powder |
Chemical Name or Material | GBR 12909 dihydrochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'