missing translation for 'onlineSavingsMsg'
Learn More
Gboxin, MedChemExpress Product Code.: 16437227

Gboxin, MedChemExpress

Product Code. 16437227
5 mg, 5mg
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16437227

Brand: Medchem Express HY111651

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Gboxin is an oxidative phosphorylation (OXPHOS) inhibitor that targets glioblastoma. Gboxin inhibits the activity of F0F1 ATP synthase. Antitumour activity.

Gboxin is an oxidative phosphorylation (OXPHOS) inhibitor that targets glioblastoma. Gboxin inhibits the activity of F0F1 ATP synthase. Antitumour activity.

Specifications

Chemical Name or Material Gboxin
CAS 2101315-36-8
Color White
Purity Grade Notes Research
Molecular Formula C22H33ClN2O2
Quantity 5 mg
Solubility Information DMSO : 32.5 mg/mL (82.71 mM; Need ultrasonic) ∣H2O : 10 mg/mL (25.45 mM; Need ultrasonic)
SMILES O=C(O[C@H]1[C@H](C(C)C)CC[C@@H](C)C1)CN2C(CC)=[N+](C)C3=CC=CC=C23.[Cl-]
Molecular Weight (g/mol) 392.96
Formula Weight 392.96
Percent Purity 99.49%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Gboxin, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.