missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
GB 31658.14-2021 Trenbolones Mixture 48 100 μg/mL in Methanol, Dr. Ehrenstorfer
Specifications
Specifications
| Kemisk navn eller materiale | 17alpha-Trenbolone∣Trenbolone |
| CAS | 80657-17-6, 10161-33-8 |
| Analyt- eller komponentnavne | 17alpha-Trenbolone∣Trenbolone |
| Forsendelsestilstand | +20°C |
| Certificeringer/compliance | ISO 17034 |
| Opløsningsmiddel eller Matrix | Methanol |
| Mængde | 1 ml |
| Koncentration eller sammensætning (efter analyt eller komponenter) | 17alpha-Trenbolone 100 μg/mL∣Trenbolone 100 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?