missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ Gastric Inhibitory Peptide, Human
For cellular and molecular biology applications
Brand: Alfa Aesar™ J66667.|D
Additional Details : Weight : 0.00001kg
Specifications
4983.6 | |
Human | |
Solid | |
RUO | |
Gastric Inhibitory Peptide, human |
HPLC | |
0.5mg | |
GIP, human | |
-30° to -10°C | |
Solid |
For Research Use Only