Få mere at vide
BOLA Gaskets for Caps
Artikelnummer. 17147434
Skift visning
Produkttype:
Septa
Artikelnummer. Produkttype
17147434 Septa
1 options
Artikelnummer. 17147434 Leverandør BOLA Leverandørnr. H97345
  • ONLINE EXCLUSIVE PRICE
    127.05 DKK
    91.08 DKK / Pakke med 5
    Spare 35.97 DKK
    28% Slukket
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
Designed specifically for use with BOLA screw caps with apertures, serving as seal or septum replacements
  • Upper side made of PTFE, lower side made of silicone-elastomer for balancing unevennesses on sealing surfaces
  • After assembly, the product is only exposed to PTFE
  • Abmessungen: For thread GL 45 Dia. of gasket 43.2 mm, Thickness of gasket 3.3 mm
Tekniske data
Farve White
Materiale PTFE-faced Silicone
Sterilitet Non-sterile
Autoklaverbar Not Autoclavable
Form Round
Produkttype Septa