Learn More
RETSCH Gasket For Grinding Jar
Brand: RETSCH 22.085.0004
1529.50 DKK valid until 2024-12-31
Use promo code "23265" to get your promotional price.
Additional Details : Weight : 0.02000kg
Specifications
For 35mL grinding jar | |
10 Pack | |
35 mL Grinding Jar, Zirconium Oxide |
Gasket | |
Zirconium Oxide |