Få mere at vide
3M™ Gas, Vapour and Particulate Filter A1B1E1K1P3 R + Formaldehyde, 6092 Artikelnummer.: 15871272
3M™ Gas, Vapour and Particulate Filter A1B1E1K1P3 R + Formaldehyde, 6092
Artikelnummer. 15871272 Shop alle 3M produkter
Filter Type: R; Formaldehyde
:
Filter Type: R; Formaldehyde
Artikelnummer. 15871272
Brand: 3M™ 7100104317_DFUOM
Se alternative produkter
3M™ Gas, Vapour and Particulate Filters, 6092 have a bayonet-style connection that fits any half and full-face 3M™ Reusable Masks, and they’re designed to optimise your field of vision. Our lightweight filters offer well-balanced ABEKP3 R breathing protection for hazardous environments.
For ABEKP3 R breathing protection against combination organic vapours (boiling point above 65°C), inorganic and acid gases, ammonia particulates and formaldehyde, use 3M™ Gas, Vapour and Particulate Filters, 6092. They’re compatible with our range of reusable half and full-face masks for safe working in hazardous environments. Our lightweight filters combine protection with comfort. They offer low breathing resistance and good balance when connected to your mask, and they’re designed to optimise your field of vision. They can be clicked into place quickly and easily, and they can be used with any half and full-face 3M™ Reusable Masks that feature a bayonet-style connection.
TRUSTED_SUSTAINABILITY
Tekniske data
Patrontype Formaldehyde
Tilslutningstype BAYONET
Filtertype R
Produkttype Combined filters
Til brug med (udstyr) Suitable for half and full-face 3M™ Reusable Masks with a bayonet-style connection
Mængde 32
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
3M™ Gas, Vapour and Particulate Filter A1B1E1K1P3 R + Formaldehyde, 6092 >