Få mere at vide
3M™ Gas, Vapour and Particulate Filter A1B1E1K1P3 R + Formaldehyde, 6092
Shop alle 3M produkter
:
Filter Type: R; Formaldehyde
Beskrivelse
For ABEKP3 R breathing protection against combination organic vapours (boiling point above 65°C), inorganic and acid gases, ammonia particulates and formaldehyde, use 3M™ Gas, Vapour and Particulate Filters, 6092. They’re compatible with our range of reusable half and full-face masks for safe working in hazardous environments. Our lightweight filters combine protection with comfort. They offer low breathing resistance and good balance when connected to your mask, and they’re designed to optimise your field of vision. They can be clicked into place quickly and easily, and they can be used with any half and full-face 3M™ Reusable Masks that feature a bayonet-style connection.
Tekniske data
Tekniske data
Patrontype | Formaldehyde |
Tilslutningstype | BAYONET |
Filtertype | R |
Produkttype | Combined filters |
Til brug med (udstyr) | Suitable for half and full-face 3M™ Reusable Masks with a bayonet-style connection |
Mængde | 32 |
Korrektion af produktindhold
Produkttitel
3M™ Gas, Vapour and Particulate Filter A1B1E1K1P3 R + Formaldehyde, 6092 >
Ser du en mulighed for forbedring?Del en indholdskorrektion