Learn More
Trajan Scientific and Medical™ Gas Tight Fixed Needle Autosampler Syringe, Side Hole Tip
Designed to provide improved solvent resistance, greater temperature range and increased operational smoothness.
Brand: Trajan Scientific and Medical™ 007891
Additional Details : Weight : 0.05000kg
Description
- High quality solutions for liquid handling
- Accurate and reproducible injections
- Specifically designed for Thermo Scientific instruments
Specifications
PTFE (Plunger) | |
0.32 mm | |
Fixed Needle | |
85 mm |
23 G | |
0.63 mm | |
500 μL | |
Side Hole |