Learn More
Trajan Scientific and Medical Gas Filter - Hydrocarbon
Prevent contaminants in gases from significantly impacting the quality of results. Oxygen, hydrocarbons and moisture can lead to problems such as noisy baselines, moisture entering the GC column, excessive bleed and septa degradation.
Brand: Trajan Scientific and Medical 1035230
Additional Details : Weight : 2.00000kg
Description
- Capacity of approximately 7 g organics dpending upon impurities, for long filter lifetime
- Can be used in combination with other filters through a simple connecting unit
Specifications
Approx. 7 g, depending upon impurities | |
Hydrocarbon | |
Connecting unit |
No | |
Removes organic compounds from gas streams |
For research use only