Beskrivelse
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human GAPDH The GAPDH Recombinant Protein Antigen is derived from E. coli. The GAPDH Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
Tekniske data
Tekniske data
| Oprensningsmetode | >80% by SDS-PAGE and Coomassie blue staining |
| Fælles navn | GAPDH Recombinant Protein Antigen |
| Indhold og opbevaring | Store at −20°C. Avoid freeze-thaw cycles. |
| Oversigt | Blocking peptide for Mfr. No. NBP2-48721 |
| Formulering | PBS and 1M Urea, pH 7.4 |
| Til brug med (applikation) | Blocking/Neutralizing, Control |
| Gene Alias | aging-associated gene 9 protein, EC 1.2.1, EC 1.2.1.12, EC 2.6.99.-, G3PD, GAPD, glyceraldehyde 3-phosphate dehydrogenase, glyceraldehyde-3-phosphate dehydrogenase, MGC88685, Peptidyl-cysteine S-nitrosylase GAPDH |
| Gen symbol | GAPDH |
| Etikettype | Unlabeled |
| Molekylvægt (g/mol) | 24 kDa |
| Vis mere |
For Research Use Only.