Få mere at vide
Novus Biologicals™ GAP-43 Recombinant Protein Antigen
Artikelnummer. 18244553 Shop alle Bio Techne produkter
Skift visning
Protein længde:
KEEPKQADVPAAVTAAAATTPAAEDAAAKATAQPPTETGESSQAEENIEAVDETKPKESARQDE
SFRGHITRKKLKGEKKDDVQAAEAEANKKDEAPVADGVEKKGEGTTTAEAAPATGSKPDEPGKAGETPSEEKKGEGDAATEQAAPQAPASSEEKAGSAETESATKA
Pakningsstørrelse:
0.1ml
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Protein længde unitSize
18244553 KEEPKQADVPAAVTAAAATTPAAEDAAAKATAQPPTETGESSQAEENIEAVDETKPKESARQDE -
18272591 SFRGHITRKKLKGEKKDDVQAAEAEANKKDEAPVADGVEKKGEGTTTAEAAPATGSKPDEPGKAGETPSEEKKGEGDAATEQAAPQAPASSEEKAGSAETESATKA 0.1ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18244553 Leverandør Novus Biologicals™ Leverandørnr. NBP254668PEP
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human GAP-43 The GAP-43 Recombinant Protein Antigen is derived from E. coli. The GAP-43 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
TRUSTED_SUSTAINABILITY
Tekniske data
Gen-id (Entrez) 2596
Protein længde KEEPKQADVPAAVTAAAATTPAAEDAAAKATAQPPTETGESSQAEENIEAVDETKPKESARQDE
Oprensningsmetode >80% by SDS-PAGE and Coomassie blue staining
Fælles navn GAP43 Recombinant Protein Antigen
Indhold og opbevaring Store at −20°C. Avoid freeze-thaw cycles.
Formulering PBS and 1M Urea, pH 7.4
Til brug med (applikation) Blocking/Neutralizing, Control
Gene Alias Axonal membrane protein GAP-43, B-50, Basp2, calmodulin-binding protein P-57, GAP43, GAP-43, growth associated protein 43, Growth-associated protein 43
Gen symbol GAP43
Etikettype Unlabeled
Molekylvægt (g/mol) 24 kDa
Produkttype Recombinant Protein Antigen
Mængde 0.1mL
Regulatorisk status RUO
Kilde E.coli
Specifik reaktivitet Human
Vis mere Vis mindre
For Research Use Only.