missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Ganglioside GM3 Antibody (racotumomab) [Janelia Fluor™ 635], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28885JF635
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Ganglioside GM3 Monoclonal antibody specifically detects Ganglioside GM3 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Tekniske data
| Ganglioside GM3 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| Human | |
| Protein A purified | |
| RUO | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Functional Assay | |
| racotumomab | |
| 50mM Sodium Borate | |
| GM3 | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion