Få mere at vide
Ganciclovir Mono-O-acetate, Mikromol™
Artikelnummer. 30459974
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30459974 4 x 25 mg
30050600 25 mg
30050184 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30459974 Leverandør Mikromol Leverandørnr. MM0485.1200254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Ganciclovir Mono-O-acetate, Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ganciclovir Mono-O-acetate
CAS 88110-89-8
Analyt- eller komponentnavne Ganciclovir Mono-O-acetate
Forsendelsestilstand Room Temperature
Molekylær formel C11 H15 N5 O5
Certificeringer/compliance ISO 9001
Synonym 6H-Purin-6-one, 9-[[2-(acetyloxy)-1-(hydroxymethyl)ethoxy]methyl]-2-amino-1,9-dihydro-, 2-[(2-Amino-1,6-dihydro-6-oxo-9H-purin-9-yl)methoxy]-3-acetoxy-1-propanol, 2-[(2-amino-6-oxo-1,6-dihydro-9H-purin-9-yl)methoxy]-3-hydroxypropyl acetate, Ganciclovir Mono-O-acetate, Monoacetoxyganciclovir, O-Acetoxyganciclovir
InChI formel InChI=1S/C11H15N5O5/c1-6(18)20-3-7(2-17)21-5-16-4-13-8-9(16)14-11(12)15-10(8)19/h4,7,17H,2-3,5H2,1H3,(H3,12,14,15,19)
Løsningstype Neat
SMIL CC(=O)OCC(CO)OCn1cnc2C(=O)NC(=Nc12)N
IUPAC navn [2-[(2-amino-6-oxo-1H-purin-9-yl)methoxy]-3-hydroxypropyl] acetate
Molekylvægt (g/mol) 297.27
Mængde 4 x 25 mg
Formel vægt 297.1073 g/mol