Få mere at vide
Ganciclovir EP Impurity J, Mikromol™
Artikelnummer. 30050324
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30050324 25 mg
30051411 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30050324 Leverandør Mikromol Leverandørnr. MM0485.100025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Ganciclovir EP Impurity J
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-[2-(Propanoylamino)-6-oxo-1,6-dihydro-9H-purin-9-yl]methoxy]propane-1,3-diyl dipropanoate (Ganciclovir Tripropionate)
CAS 177216-32-9
Analyt- eller komponentnavne 2-[2-(Propanoylamino)-6-oxo-1,6-dihydro-9H-purin-9-yl]methoxy]propane-1,3-diyl dipropanoate (Ganciclovir Tripropionate)
Forsendelsestilstand Room Temperature
Molekylær formel C18 H25 N5 O7
Certificeringer/compliance ISO 17025
Synonym 2-[2-(Propanoylamino)-6-oxo-1,6-dihydro-9H-purin-9-yl]methoxy]propane-1,3-diyl dipropanoate, Ganciclovir Imp. J (EP), Ganciclovir Tripropionate, Ganciclovir Impurity J
InChI formel InChI=1S/C18H25N5O7/c1-4-12(24)20-18-21-16-15(17(27)22-18)19-9-23(16)10-30-11(7-28-13(25)5-2)8-29-14(26)6-3/h9,11H,4-8,10H2,1-3H3,(H2,20,21,22,24,27)
SMIL CCC(=O)NC1=Nc2c(ncn2COC(COC(=O)CC)COC(=O)CC)C(=O)N1
IUPAC navn [2-[[6-oxo-2-(propanoylamino)-1H-purin-9-yl]methoxy]-3-propanoyloxypropyl] propanoate
Molekylvægt (g/mol) 423.42
Mængde 25 mg
Formel vægt 423.1754 g/mol