Få mere at vide
Ansell™ Gammex™ PF Latex Surgical Gloves
Artikelnummer. 12302078 Shop alle Ansell produkter
Encompass
Skift visning
Pakningsstørrelse:
Pakke med 40
Artikelnummer. unitSize
12302078 Pakke med 40
1 options
Artikelnummer. 12302078 Leverandør Ansell™ Leverandørnr. 351144
Ansell™ Gammex™ PF Latex Surgical Gloves have low allergenicity and provide a secure grip on instruments for easy, sure handling in both wet and dry conditions.
  • Superb grip;and handling with micro-rough outer surface
  • Very high tear resistance
  • Low allergenicity - the Gammex™ PF glove reduces possible contact with potential latex allergens thanks to its synthetic inner coating which provides a protective shield
  • Resistant to alcohol-based hand disinfectants
  • Resistant to viral penetration as verified by extensive bacteriophage test results
Tekniske data
Certificeringer/compliance CE Class Iia; CE mark; EN 455 parts 1,2 and 3; ISO 9001 and/or ISO 13485
Belægningsmateriale Polymer coated and siliconised polyurethane coating
Farve White
Engangs Single-use
Udvendig finish Micro Textured
Hånd Ambidextrous
Latex fri No
Materiale Latex
Pulveriseret Powder Free
Produkttype Latex surgical gloves
Sterilitet Sterile
Tykkelse (metrisk) 0.2 mm
Handske manchet stil Straight cuff with adhesive band
Længde (metrisk) 290 mm