Få mere at vide
Gamma-Oxo-3-pyridinebutyric Acid, TRC
Artikelnummer. 30136746
Skift visning
Mængde:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136746 25 mg
30009523 10 mg
30004206 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30136746 Leverandør Toronto Research Chemicals Leverandørnr. TRCO85916025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 4192-31-8
Molekylær formel C9 H9 N O3
Molekylvægt (g/mol) 179.17
Synonym γ-Oxo-3-pyridinebutanoic Acid, 3-Succinoylpyridine, 4-(3-Pyridyl)-4-oxobutyric Acid, OPBA
IUPAC navn 4-oxo-4-(3-pyridyl)butanoic acid
SMIL OC(=O)CCC(=O)c1cccnc1
Tekniske data
Kemisk navn eller materiale gamma-Oxo-3-pyridinebutyric Acid
Mængde 25 mg
InChI formel InChI=1S/C9H9NO3/c11-8(3-4-9(12)13)7-2-1-5-10-6-7/h1-2,5-6H,3-4H2,(H,12,13)
Formel vægt 179.058