Få mere at vide
Gamma-Hydroxy Phenylbutazone, TRC
Artikelnummer. 30136068
Skift visning
Mængde:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136068 25 mg
30134789 5 mg
30007346 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30136068 Leverandør Toronto Research Chemicals Leverandørnr. TRCH94007025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 568-76-3
Molekylær formel C19 H20 N2 O3
Molekylvægt (g/mol) 324.37
Synonym 4-(3-Hydroxybutyl)-1,2-diphenyl-3,5-pyrazolidinedione;
IUPAC navn 4-(3-hydroxybutyl)-1,2-diphenylpyrazolidine-3,5-dione
SMIL CC(O)CCC1C(=O)N(N(C1=O)c2ccccc2)c3ccccc3
Tekniske data
Kemisk navn eller materiale gamma-Hydroxy Phenylbutazone
Mængde 25 mg
InChI formel InChI=1S/C19H20N2O3/c1-14(22)12-13-17-18(23)20(15-8-4-2-5-9-15)21(19(17)24)16-10-6-3-7-11-16/h2-11,14,17,22H,12-13H2,1H3
Formel vægt 324.1474