Få mere at vide
Gamma-Butyrobetaine-d9, TRC
Artikelnummer. 30345789
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30345789 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30345789 Leverandør Toronto Research Chemicals Leverandørnr. TRCB75949750MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale gamma-Butyrobetaine trimethyl-d9
CAS 479677-53-7
Molekylær formel C7 2H9 H7 N O2
Mængde 50 mg
Synonym 3-Carboxy-N,N,N-tri(methyl-d3)-1-propanaminium, Actinine-d9, 4-Butyrobetaine-d9, Butyrobetaine-d9, Deoxycarnitine-d9, γ-Aminobutyric-d9 Acid Betaine, γ-Butyrobetaine-d9
InChI formel InChI=1S/C7H15NO2/c1-8(2,3)6-4-5-7(9)10/h4-6H2,1-3H3/p+1/i1D3,2D3,3D3
SMIL [2H]C([2H])([2H])[N+](CCCC(=O)O)(C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC navn 3-carboxypropyl-tris(trideuteriomethyl)azanium
Molekylvægt (g/mol) 155.26
Formel vægt 155.1746