Få mere at vide
Gamma-Butyrobetaine-d9 Hydrochloride, TRC
Artikelnummer. 30347089
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30347089 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30347089 Leverandør Toronto Research Chemicals Leverandørnr. TRCB75950250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale gamma-Butyrobetaine trimethyl-d9 hydrochloride
CAS 85806-17-3
Molekylær formel C7 2H9 H7 N O2 . Cl
Mængde 50 mg
Synonym 1-Propanaminium, 3-carboxy-N,N,N-tri(methyl-d3)-, chloride (1:1) (ACI), 1-Propanaminium, 3-carboxy-N,N,N-tri(methyl-d3)-, chloride (9CI), 3-Carboxy-N,N,N-tri(methyl-d3)-1-propanaminium chloride, N-(Carboxypropyl)-N,N,N-trimethyl-d9-ammonium Chloride, γ-Butyrobetaine-d9 Hydrochloride, (3-Carboxypropyl)trimethylammonium-d9 Chloride, (3-Carboxypropyl)trimethylammonium-D9 (N,N,N-trimethyl-D9) Chloride
InChI formel InChI=1S/C7H15NO2.ClH/c1-8(2,3)6-4-5-7(9)10;/h4-6H2,1-3H3;1H/i1D3,2D3,3D3;
SMIL [Cl-].[2H]C([2H])([2H])[N+](CCCC(=O)O)(C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC navn 3-carboxypropyl-tris(trideuteriomethyl)azanium;chloride
Molekylvægt (g/mol) 190.72
Formel vægt 190.1434