Få mere at vide
gamma-1,2,3,4,5,6-Hexachlorocyclohexane-d6, CDN
Artikelnummer. 30328409
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30328409 0.01 g
30323011 0.005 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30328409 Leverandør CDN Leverandørnr. CDND30100.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale gamma-HCH D6
CAS 60556-82-3
Molekylær formel C6 2H6 Cl6
Mængde 0.01 g
Synonym (1α,2α,3β,4α,5α,6β)-1,2,3,4,5,6-Hexachlorocyclohexane-1,2,3,4,5,6-d6, Hexadeuterated lindane, Lindane-d6, gamma-HCH D6
InChI formel InChI=1 S/C6H6Cl6/c7-1-2(8)4(10)6(12)5(11)3(1)9/h1-6 H/t1-,2-,3-,4+,5+,6+/i1D,2 D,3 D,4 D,5 D,6 D
SMIL [2 H][C@@]1(Cl)[C@@]([2 H])(Cl)[C@@]([2 H])(Cl)[C@]([2 H])(Cl)[C@@]([2 H])(Cl)[C@@]1([2 H])Cl
IUPAC navn 1,2,3,4,5,6-hexachloro-1,2,3,4,5,6-hexadeuteriocyclohexane
Molekylvægt (g/mol) 296.87
Formel vægt 293.8977 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity