Få mere at vide
WTW™ Galvanisk opløst iltsensor til WTW Ledningsevne og multiparameter
Beskrivelse
The CellOx™ 325 is a dissolved oxygen sensor with integrated temperature compensation and membrane leak monitoring, which can be used in the lab as well as in the field. Its features include a quick response time (t99<60 s) and low-maintenance operation. Its fields of use include surface waters, measurements in wastewater treatment plants, BOD directions, but also industrial and other applications. It is filled with electrolyte and achieves a long dwelling time up to 6 months. The calibration vessel is part of the delivery.
Tekniske data
Tekniske data
| Oversigt | CellOx™ 325-20 |
| Længde (metrisk) kabel | 20 m |
| Inkluderer | 20m L Cable Cleaning solution, abrasive foil, 3 exchange membrane heads and electrolyte |
| Til brug med (udstyr) | Conductivity and multiparameter meters |
| Model- | CellOx 325 |