missing translation for 'onlineSavingsMsg'
Få mere at vide

Thermo Scientific™ Gallery™ System Reagents for Environmental and Industrial Analysis

Artikelnummer. 15760339 Shop alle Thermo Scientific produkter
Skift visning
Klik for at se tilgængelige muligheder
Type:
Fosfatsystemreagens
Antal prøver:
2300
Pakningsstørrelse:
20ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Type Antal prøver unitSize
15760339 Fosfatsystemreagens 2300 20ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Artikelnummer. 15760339 Leverandør Thermo Scientific™ Leverandørnr. 984366

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

Denne vare kan ikke returneres. Se returpolicy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Select from these ready-to-use reagents and enzymatic kits specifically designed for environmental and industrial water analysis using a Thermo Scientific™ Gallery™ Discrete Analyzer.

Save time and reduce errors by eliminating reagent preparation with the Thermo Scientific™ Gallery™ system reagents for environmental testing (clean water, wastewater, soil and sludge digests, effluents, and saline) and industrial (process water) analysis. With over 40 ready-to-use Gallery system reagents, these kits include 1 to 3 reagents in 20 mL barcoded vials, allowing the necessary identification data, such as lot and expiration date, to be read automatically by the Gallery Discrete Analyzer's barcode reader. Automated dilutions make it possible to apply methods for a wide range of analyte concentrations and the test applications are based on recommended standard water and wastewater methods used in most countries.

Highlights:
  • All test applications are based on recommended standard methods in water and wastewater analysis of different countries
  • Kit structure includes 1 to 3 reagents in 20mL vials
  • Barcoded vials are simple and fast to use
  • All necessary data, such as lot and expiration date, are automatically read
  • Automated dilutions make it possible to apply methods for a wide range of analyte concentrations
  • Types of matrices analyzed include clean water, wastewater, soil and sludge digests, effluents, saline and process water
TRUSTED_SUSTAINABILITY

Tekniske data

Type Fosfatsystemreagens
Prøvetype Water Sample, Soil Sample
Oversigt Phosphate system reagents - R1 (2300 tests)
Registrerer Phosphate R1
Antal prøver 2300
Testområde 1.0 to 10.0
Mængde 4 x 20 mL
Interferensfilter 880
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.