Få mere at vide
Gallamine Triethiodide, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Gallamine triethiodide |
| CAS | 65-29-2 |
| Molekylær formel | C30 H60 N3 O3 . 3 I |
| Mængde | 5 g |
| Synonym | Ammonium, [v-phenenyltris(oxyethylene)]tris[triethyl-, triiodide (8CI), Ethanaminium, 2,2',2''-[1,2,3-benzenetriyltris(oxy)]tris[N,N,N-triethyl-, triiodide (9CI), [v-Phenenyltris(oxyethylene)]tris[triethylammonium iodide] (6CI,7CI), 1,2,3-Tri(β-diethylaminoethoxy)benzene triethiodide, 1,2,3-Tris(2-diethylaminoethoxy)benzene triethiodide, 1,2,3-Tris(2-diethylaminoethoxy)benzene tris(ethyliodide), 1,2,3-Tris(2-triethylammonium ethoxy)benzene triiodide, 1,2,3-Tris(diethylaminoethoxy)benzene triethiodide, 3697 RP, Benzcurine iodide, F 2559, Flacedil, Flaxedil, Flaxedil iodide, Fourneau 2559, Gallaflex, Gallamin triethiodide, Gallamine iodide, Gallamine triethiodide, Gallamine triiodoethylate, HL 8583, Miowas G, Parexyl, Pyrogallol 1,2,3-(diethylaminoethyl ether) tris(ethyliodide), Pyrolaxon, RP 3697, Relaxan, Remyolan, Retensin, Sincurarine, Syncurarine, Tri(diethylaminoethoxy)-1,2,3-benzene triiodoethylate, Tri(β-diethylaminoethoxy)-1,2,3-benzene tri-iodoethylate, Ethanaminium, 2,2',2''-[1,2,3-benzenetriyltris(oxy)]tris[N,N,N-triethyl-, iodide (1:3) |
| InChI formel | InChI=1S/C30H60N3O3.3HI/c1-10-31(11-2,12-3)22-25-34-28-20-19-21-29(35-26-23-32(13-4,14-5)15-6)30(28)36-27-24-33(16-7,17-8)18-9;;;/h19-21H,10-18,22-27H2,1-9H3;3*1H/q+3;;;/p-3 |
| SMIL | [I-].[I-].[I-].CC[N+](CC)(CC)CCOc1cccc(OCC[N+](CC)(CC)CC)c1OCC[N+](CC)(CC)CC |
| IUPAC navn | 2-[2,3-bis[2-(triethylazaniumyl)ethoxy]phenoxy]ethyl-triethylazanium;triiodide |
| Molekylvægt (g/mol) | 891.53 |
| Formel vægt | 891.1769 |