Få mere at vide
Tocris Bioscience™ Galeterone, Tocris Bioscience™
CYP17 inhibitor; also androgen receptor antagonist
Brand: Tocris Bioscience™ 6108/50
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Galeterone is a CYP17 inhibitor (IC50 = 300 nM) and androgen receptor antagonist (IC50 = 384 nM in PC3-AR cells). Inhibits DHT-stimulated prostate cancer cell proliferation in vitro. Exhibits antitumor activity in SCID mice bearing prostate cancer xenografts.Tekniske data
| Galeterone | |
| (3β)-17-(1H-Benzimidazol-1-yl)androsta-5,16-dien-3-ol | |
| Soluble to 50mM in DMSO and to 50mM in ethanol | |
| White |
| 851983-85-2 | |
| 50 mg | |
| Solid | |
| Bioactive Small Molecule |
Produkttitel
Ser du en mulighed for forbedring?
For Research Use Only.