Få mere at vide
Tocris Bioscience™ Galeterone
GREENER_CHOICE
Artikelnummer. 18799607
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18799607 10 mg
18718138 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18799607 Leverandør Tocris Bioscience™ Leverandørnr. 6108/10
CYP17 inhibitor; also androgen receptor antagonist
Galeterone is a CYP17 inhibitor (IC50 = 300 nM) and androgen receptor antagonist (IC50 = 384 nM in PC3-AR cells). Inhibits DHT-stimulated prostate cancer cell proliferation in vitro. Exhibits antitumor activity in SCID mice bearing prostate cancer xenografts.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Galeterone
CAS 851983-85-2
Synonym (3β)-17-(1H-Benzimidazol-1-yl)androsta-5,16-dien-3-ol
Mængde 10 mg
Opløselighed Soluble to 50mM in DMSO and to 50mM in ethanol
Molekylær formel C26H32N2O
Formel vægt 388.55
Fysisk form Solid
Farve White
Produkttype Bioactive Small Molecule
Renhed >98%
Vis mere Vis mindre
For Research Use Only.