Få mere at vide
Eppendorf™ Galaxy™ 48 R
Artikelnummer. 15431394
Inkluderer:
High Temp Disinfection
No High Temp Disinfection
Pakningsstørrelse:
Hver
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.
Artikelnummer. 15431394
Brand: Eppendorf™ CO48330001
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.
Galaxy™ 48 R is ideally suited for applications requiring a high level of cell culture isolation.
  • Provides mid-sized capacity within a small footprint
  • Fanless design offers more usable space
  • Six-sided direct heating system provides a uniform incubated environment
  • Deep-drawn, easy-to-clean stainless steel chamber without seams or welds
  • Low gas consumption
  • Integrated alarm relay
TRUSTED_SUSTAINABILITY
Tekniske data
Kapacitet (metrisk) 48 L
Ilt kontrol 1 to 19 %
Temperaturområde (metrisk) Ambient +4° to 50°C
Vise Standard display
Inkluderer High Temp Disinfection
Havne 1
Mål (D x B x H) Indvendig 30.5 x 401 x 40.1 cm
Dimensioner (L x B x H) 47.5 x 48.3 x 64.5 cm