Få mere at vide
Galanthamine-O-methyl-d3, TRC
Artikelnummer. 30240595
Skift visning
Mængde:
2.5 mg
25 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30240595 2.5 mg 2.5mg
30240479 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30240595 Leverandør Toronto Research Chemicals Leverandørnr. TRCG1885022.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1279031-09-2
Molekylær formel C17 2H3 H18 N O3
Molekylvægt (g/mol) 290.37
Synonym 6H-Benzofuro[3a,3,2-ef][2]benzazepin-6-ol, 4a,5,9,10,11,12-hexahydro-3-(methoxy-d3)-11-methyl-, (4aS,6R,8aS)-, (4aS,6R,8aS)-4a,5,9,10,11,12-Hexahydro-3-(methoxy-d3)-11-methyl-6H-benzofuro[3a,3,2-ef][2]benzazepin-6-ol, Galanthamine-O-methyl-d3, Galantamine D3 (O-methyl D3)
SMIL [2H]C([2H])([2H])Oc1ccc2CN(C)CC[C@@]34C=C[C@H](O)C[C@@H]3Oc1c24
Tekniske data
Kemisk navn eller materiale Galanthamine-O-methyl-D3
Renhedsgrad noter HPLC
Mængde 2.5 mg
InChI formel InChI=1S/C17H21NO3/c1-18-8-7-17-6-5-12(19)9-14(17)21-16-13(20-2)4-3-11(10-18)15(16)17/h3-6,12,14,19H,7-10H2,1-2H3/t12-,14-,17-/m0/s1/i2D3
Formel vægt 290.171
Procent renhed >95%