Få mere at vide
Galanthamine-O-(methyl-d3)-N-(methyl-d3), TRC
Artikelnummer. 30240916
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30240916 1 mg
30240989 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30240916 Leverandør Toronto Research Chemicals Leverandørnr. TRCG1885031MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1128109-00-1
Molekylær formel C17H15D6NO3
Molekylvægt (g/mol) 293.39
SMIL [2H]C([2H])([2H])Oc1ccc2CN(CC[C@@]34C=C[C@H](O)C[C@@H]3Oc1c24)C([2H])([2H])[2H]
Tekniske data
Kemisk navn eller materiale Galanthamine-O-(methyl-d3)-N-(methyl-d3)
Mængde 1 mg
InChI formel InChI=1S/C17H21NO3/c1-18-8-7-17-6-5-12(19)9-14(17)21-16-13(20-2)4-3-11(10-18)15(16)17/h3-6,12,14,19H,7-10H2,1-2H3/t12-,14-,17-/m0/s1/i1D3,2D3
Formel vægt 293.1898