missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Galanin R2/GALR2 Antibody (616520) [Biotin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB6544B
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Galanin R2/GALR2 Monoclonal antibody specifically detects Galanin R2/GALR2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF
Tekniske data
| Galanin R2/GALR2 | |
| Monoclonal | |
| Biotin | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry, CyTOF | |
| 616520 | |
| Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
| GAL2-R, galanin receptor 2, galanin receptor type 2, GALNR2MGC125984, GALR-2, MGC125983 | |
| NS0 mouse myeloma cell line transfected with human Galanin R2, Accession # O43603 | |
| 0.1 mL | |
| GPCR | |
| 8811 | |
| Store at 4°C in the dark. | |
| IgG2b |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion