Få mere at vide
Gadopentetate dimeglumine, MedChemExpress
Artikelnummer. 16461356
Encompass_Preferred
Skift visning
Mængde:
50 mg
100 mg
200 mg
500 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16461356 200 mg
16498036 100 mg
16441986 500 mg
16434197 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 16461356 Leverandør Medchem Express Leverandørnr. HYA0167
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
MedChemExpress Gadopentetate dimeglumine (SH-L-451A) is used in combination with magnetic resonance imaging (MRI) to allow blood vessels, organs, and other non-bony tissues to be seen more clearly on the MRI.
Gadopentetate dimeglumine (SH-L-451A) is used in combination with magnetic resonance imaging (MRI) to allow blood vessels, organs, and other non-bony tissues to be seen more clearly on the MRI.
Tekniske data
Kemisk navn eller materiale Gadopentetate dimeglumine
Fysisk form Solid
CAS 86050-77-3
Farve White
Mængde 200 mg
Renhedsgrad noter Research
Molekylær formel C28H54GdN5O20
Til brug med (applikation) Cancer-programmed cell death
Synonym SH-L-451A Gadopentetic acid dimeglumine salt
Opløselighedsinformation DMSO : 100 mg/mL (106.61 mM; Need ultrasonic)
SMIL O=C1[O-][Gd+3]([N]2(C3)CC4=O)([N]5(C6)CC7=O)([O-]C6=O)([O-]7)([O-]C3=O)([O-]4)[N](CC5)(CC2)C1.O[C@H]([C@@H](O)CNC)[C@H](O)[C@H](O)CO.O[C@H]([C@@H](O)CNC)[C@H](O)[C@H](O)CO.[2H+]
Molekylvægt (g/mol) 938.0
Formel vægt 938.0
Procent renhed 99.0%