missing translation for 'onlineSavingsMsg'
Learn More

Gadolinium foil, 0.25mm (0.01in) thick, 99.9% (REO), Thermo Scientific Chemicals

Catalog Number p-7048993
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
This item is not returnable. View return policy
7440-54-2
Gd
157.25
MFCD00011022
UIWYJDYFSGRHKR-UHFFFAOYSA-N
atom, gadolinio, unii-au0v1lm3jt, trihydride, au0v1lm3jt, foil, 3n, gadolinium, elemental, ingot, gadolinium, chips, compounds
23982
CHEBI:33375
gadolinium
[Gd]
This item is not returnable. View return policy
7440-54-2
Gd
157.25
MFCD00011022
UIWYJDYFSGRHKR-UHFFFAOYSA-N
atom, gadolinio, unii-au0v1lm3jt, trihydride, au0v1lm3jt, foil, 3n, gadolinium, elemental, ingot, gadolinium, chips, compounds
23982
CHEBI:33375
gadolinium
[Gd]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY
UN Number UN3178
Quantity 25 x 25 mm
Percent Purity 99.9%
Weight ≈1.2g/25x25mm
Form Foil
Assay (REO)
Chemical Name or Material Gadolinium foil, 0.25mm (0.01 in.) thick

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'