missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ GAD65 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA521297
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Recommended positive controls: Mouse brain, rat brain. Predicted reactivity: Mouse (100%), Rhesus Monkey (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
GAD65 (glutamic acid decarboxylase-65) are present in autoimmune disorders such as insulin-dependent (type1) diabetes mellitus (IDDM), stiff man syndrome and polyendocrine autoimmune disease. Auto-antibodies to GAD65 are present in 60-70% of individuals with newly diagnosed IDDM and thus are important markers for disease activity. These auto-antibodies usually recognize conformation-dependent regions on GAD65 and rarely bind to the second isoform, GAD67. Auto-antibodies to GAD67 are found in only 15% of recent-onset IDDM patients and most of this binding can be blocked with GAD65, suggesting shared epitopes between the two isoforms of GAD.
Tekniske data
| GAD65 | |
| Polyclonal | |
| Unconjugated | |
| GAD2 | |
| 6330404F12Rik; 65 kDa glutamic acid decarboxylase; GAD(65); GAD2; Gad-2; GAD65; GAD-65; Glutamate decarboxylase 2; Glutamate decarboxylase 2 (islet); glutamate decarboxylase 2 (pancreatic islets and brain, 65kDa); glutamate decarboxylase 65 kDa isoform; Glutamate decarboxylase-2 (pancreas); glutamic acid decarboxylase 2; glutamic acid decarboxylase 65; MGC161605; MGC161607; RP11-420F12.2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 14417, 24380, 2572 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
| P48320, Q05329, Q05683 | |
| GAD2 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human GAD65. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion