missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
GAD1/GAD67 Antibody (GAD1/2563), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79937PCP
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
GAD1/GAD67 Monoclonal specifically detects GAD1/GAD67 in Human samples. It is validated for Flow Cytometry.
Tekniske data
| GAD1/GAD67 | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| GAD1 | |
| Recombinant human GAD1/GAD67 protein fragment (around aa 72-135) (exact sequence is proprietary) (Uniprot: Q99259) | |
| 0.1 mL | |
| Diabetes Research | |
| This monoclonal antibody recognizes a protein of 67kDa, which is identified as glutamic acid decarboxylase 1 (GDA1). There are two forms of glutamic acid decarboxylases (GADs) that are found in the brain: GAD65 (also known as GAD2) and GAD67 (also known as GAD1. GAD65 and GAD67 are members of the group II decarboxylase family of proteins and are responsible for catalyzing the rate-limiting step in the production of GABA (-aminobutyric acid) from L-glutamic acid. Although both GADs are found in the brain, GAD65 localizes to synaptic vesicle membranes in nerve terminals, while GAD67 is distributed throughout the cell. GAD67 is responsible for the basal levels of GABA synthesis. In the case of a heightened demand for GABA in neurotransmission, GAD65 will transiently activate to assist in GABA production. The loss of GAD65 is detrimental and can impair GABA neurotransmission, however the loss of GAD67 is lethal. | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| GAD1/2563 | |
| Flow Cytometry | |
| 67kD), EC 4.1.1, FLJ45882, GAD25, glutamate decarboxylase 1 (brain, 67kDa), Glutamate decarboxylase 67 kDa isoform | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| IgG2b κ |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion