Få mere at vide
Gabapentin Enacarbil Sodium Salt, TRC
Artikelnummer. 30135660
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30135660 250 mg
30007184 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30135660 Leverandør Toronto Research Chemicals Leverandørnr. TRCG117240250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 912486-72-7
Molekylær formel C16 H26 N O6 . Na
Molekylvægt (g/mol) 351.37
Synonym 1-[[(1-(Isobutanoyloxy)ethoxy)carbonyl]aminomethyl]cyclohex-1-ylacetic Acid Sodium Salt, Horizant Sodium Salt, Regnite Sodium Salt, XP 13512 Sodium Salt
IUPAC navn sodium;2-[1-[[1-(2-methylpropanoyloxy)ethoxycarbonylamino]methyl]cyclohexyl]acetate
SMIL [Na+].CC(C)C(=O)OC(C)OC(=O)NCC1(CC(=O)[O-])CCCCC1
Tekniske data
Kemisk navn eller materiale Gabapentin Enacarbil sodium salt
InChI formel InChI=1S/C16H27NO6.Na/c1-11(2)14(20)22-12(3)23-15(21)17-10-16(9-13(18)19)7-5-4-6-8-16;/h11-12H,4-10H2,1-3H3,(H,17,21)(H,18,19);/q;+1/p-1
Mængde 250 mg
Formel vægt 351.1658