Få mere at vide
G 5555, Tocris Bioscience™
High affinity group I PAK inhibitor
Brand: Tocris Bioscience 6051/50
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
G 5555 is a high affinity group 1 PAK (pan-PAK1, 2 and 3) inhibitor (Ki = 3.7 nM; pMEK IC50 = 69 nM). Of 235 kinases tested, other than PAK1, eight (PAK2, PAK3, KHS1, Lck, MST3, MST4, SIK2, and YSK1) were inhibited >70%. Exhibits low hERG channel activity (<50% inhibition at 10 μM). Orally bioavailable.Tekniske data
| 8-[(trans-5-Amino-1,3-dioxan-2-yl)methyl]-6-[2-chloro-4-(6-methyl-2-pyridinyl)phenyl]-2-(methylamino)-pyrido[2,3-d]pyrimidin-7(8H)-one | |
| 1648863-90-4 | |
| >98% | |
| 50 mg |