missing translation for 'onlineSavingsMsg'
Få mere at vide

Tocris Bioscience™ G-1

Artikelnummer. 18094882
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18094882 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 18094882 Leverandør Tocris Bioscience™ Leverandørnr. 3577/10

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Potent and selective GPER agonist

G-1 is a potent and selective GPER agonist (Ki = 11 nM, EC50 = 2 nM); displays no activity at ERα and ERβ at concentrations up to 10 μM. Increases cytosolic Ca2+ and inhibits migration of SKBr3 cells and MCF-7 cells in response to chemoattractants (IC50 values are 0.7 and 1.6 nM respectively) in vitro. Blocks MCF-1 cell cycle progression at the G1 phase. Displays therapeutic effects in the mouse EAE model of multiple sclerosis. Also inhibits glutamate-induced autophagy and neuronal loss in cultured primary cortical neurons.
TRUSTED_SUSTAINABILITY

Specifica

Kemisk navn eller materiale (±)-1-[(3aR*,4S*,9bS*)-4-(6-Bromo-1,3-benzodioxol-5-yl)-3a,4,5,9b-tetrahydro-3H-cyclopenta[c]quinolin-8-yl]- ethanone
CAS 881639-98-1
Mængde 10 mg
Mål Estrogen and Related Receptor Agonists
Molekylær formel C21H18BrNO3
Renhed 98%
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.