missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human FYN A, GST Tag Recombinant Protein

Human FYN A, GST Tag Recombinant Protein

Brand:  Invitrogen™ PV6346

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15428983

  • 3660.00 DKK / 10µg
Estimated Shipment: 14-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Fyn has been shown to phosphorylate Dab1, an intracellular adaptor protein that interacts with amyloid precursor protein (APP) and apoE receptor 2 (apoEr2). The interaction of Fyn and Dab1 regulates the phosphorylation, trafficking, and processing of APP and apoEr2. Fyn expression has been shown to be significantly increased in Chronic Myelogenous Leukemia (CML). Knockdown of Fyn with shRNA slows leukemia cell growth, inhibits clonogenicity, and leads to increased sensitivity to imatinib.

  • Validated in LanthaScreen™ Kinase Binding Assay
  • Protocols available for simple use and optimized performance
  • Multiple pack sizes available for convenient use
Specifications

Specifications

Enzyme and Protein Activity Assay, Industrial and Applied Science, Kinase Assay, Kinase Biology, Kinase Proteins and Expression Reagents, Pharma and Biopharma, Protein Assays and Analysis, Protein Biology, Target and Lead Identification and Validation
87.1kDa
Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws.
Human
GST Tag
50mM Tris-HCl pH 7.5, 150mM NaCl, 0.25mM DTT, 10mM glutathione, 0.1mM EDTA, 0.1mM PMSF, 25% glycerol
10μg
SLK, SYN, MGC45350
Recombinant
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers