missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Tocris Bioscience™ FX 1

Description
FX 1 is a Bcl-6 inhibitor (Kd = 3 μM); exhibits 10-fold greater potency than endogenous Bcl-6 corepressors. Disrupts formation of Bcl-6 repression complex. Induces regression of established tumors in mice bearing DLBCL xenograft.
Specifications
Specifications
| Kemisk navn eller materiale | (5Z)-5-(5-Chloro-1,2-dihydro-2-oxo-3H-indol-3-ylidene)-4-oxo-2-thioxo-3-thiazolidinepropanoic acid |
| CAS | 1426138-42-2 |
| Mængde | 25 mg |
| Molekylær formel | C14H9ClN2O4S2 |
| Formel vægt | 368.82 |
| Renhed | 98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?