Få mere at vide
Fusidic Acid Hemihydrate, LoGiCal
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Fusidic acid hemihydrate |
| CAS | 172343-30-5 |
| Analyt- eller komponentnavne | Fusidic acid hemihydrate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | 2 C31 H48 O6 . H2 O |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Fenoprofen 1,3-Butylene Glycol Esters (Mixture of Regio- and Stereoisomers), ent-(17Z)-16alpha-(Acetyloxy)-3beta,11beta-dihydroxy-4beta,8,14-trimethyl-18-nor-5beta,10alpha-cholesta-17(20),24-dien-21-oic acid hemihydrate, 29-Nordammara-17(20),24-dien-21-oic acid, 16-(acetyloxy)-3,11-dihydroxy-, hydrate (2:1), (3alpha,4alpha,8alpha,9beta,11alpha,13alpha,14beta,16beta,17Z)-, Fusidic acid hemihydrate |
| InChI formel | InChI=1S/2C31H48O6.H2O/c2*1-17(2)9-8-10-20(28(35)36)26-22-15-24(34)27-29(5)13-12-23(33)18(3)21(29)11-14-30(27,6)31(22,7)16-25(26)37-19(4)32;/h2*9,18,21-25,27,33-34H,8,10-16H2,1-7H3,(H,35,36);1H2/b2*26-20-;/t2*18-,21-,22-,23+,24+,25-,27-,29-,30-,31-;/m00./s1 |
| Løsningstype | Neat |
| SMIL | O.C[C@@H]1[C@H](O)CC[C@@]2(C)[C@H]1CC[C@@]3(C)[C@H]2[C@H](O)C[C@H]4\C(=C(/CCC=C(C)C)\C(=O)O)\[C@H](C[C@]34C)OC(=O)C.C[C@@H]5[C@H](O)CC[C@@]6(C)[C@H]5CC[C@@]7(C)[C@H]6[C@H](O)C[C@H]8\C(=C(/CCC=C(C)C)\C(=O)O)\[C@H](C[C@]78C)OC(=O)C |
| Vis mere |