missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
FUS Antibody (CL0190) [mFluor Violet 610 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52874MFV610
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
FUS Monoclonal antibody specifically detects FUS in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Gene Knock-Out,KnockDown,Immunocytochemistry/ Immunofluorescence
Tekniske data
| FUS | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 75 kDa DNA-pairing protein, ALS6, amyotrophic lateral sclerosis 6, FUS1, fused in sarcoma, fusion (involved in t(12;16) in malignant liposarcoma), fusion gene in myxoid liposarcoma, fusion, derived from t(12;16) malignant liposarcoma, fus-like protein, heterogeneous nuclear ribonucleoprotein P2, HNRNPP2, Oncogene FUS, Oncogene TLS, POMp75, RNA-binding protein FUS, TLShnRNP-P2, Translocated in liposarcoma protein | |
| This antibody was developed using a recombinant protein derived from P35637, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer, Neuroscience | |
| 2521 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Gene Knock-Out, KnockDown, Immunocytochemistry/Immunofluorescence | |
| CL0190 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion