missing translation for 'onlineSavingsMsg'
Learn More

Furfuryl isothiocyanate, 97%, Thermo Scientific Chemicals

Catalog Number p-7024552
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
4650-60-6
C6H5NOS
139.172
MFCD00014112
ICXYINJACKJQQV-UHFFFAOYSA-N
furfuryl isothiocyanate, 2-isothiocyanatomethyl furan, 2-furylmethyl isothiocyanate, 2-isothiocyanatomethyl-furan, furan, 2-isothiocyanatomethyl, 2-furanylmethyl isothiocyanate, 2-furylmethanisothiocyanate, furfurylisothiocyanate, furylmethylisothiocyanate, 7-bromo-quinolin-2-ylamine
78380
2-(isothiocyanatomethyl)furan
C1=COC(=C1)CN=C=S
This item is not returnable. View return policy
4650-60-6
C6H5NOS
139.172
MFCD00014112
ICXYINJACKJQQV-UHFFFAOYSA-N
furfuryl isothiocyanate, 2-isothiocyanatomethyl furan, 2-furylmethyl isothiocyanate, 2-isothiocyanatomethyl-furan, furan, 2-isothiocyanatomethyl, 2-furanylmethyl isothiocyanate, 2-furylmethanisothiocyanate, furfurylisothiocyanate, furylmethylisothiocyanate, 7-bromo-quinolin-2-ylamine
78380
2-(isothiocyanatomethyl)furan
C1=COC(=C1)CN=C=S

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.39
Boiling Point 64°C to 68°C (2 mmHg)
Odor Pungent
Refractive Index 1.5655
Quantity 5 g
UN Number UN2922
Beilstein 1281561
Sensitivity Moisture sensitive
Formula Weight 139.18
Percent Purity 97%
Chemical Name or Material Furfuryl isothiocyanate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'