Få mere at vide
Furfuryl-d5-amine, CDN
Artikelnummer. 30323516
Skift visning
Mængde:
0.01 g
0.05 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30323516 0.05 g
30319835 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30323516 Leverandør CDN Leverandørnr. CDND65020.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Furfurylamine-D5
CAS 1219799-26-4
Molekylær formel C5 D5 H2 N O
Mængde 0.05 g
Synonym Furfuryl-d5-amine, (Furan-2-yl-3,4,5-d3)methan-d2-amine, 2-Furanylmethanamine-d5, 2-Furfurylamine-D5
InChI formel InChI=1 S/C5H7NO/c6-4-5-2-1-3-7-5/h1-3 H,4,6H2/i1D,2 D,3 D,4D2
SMIL [2 H]c1oc(c([2 H])c1[2 H])C([2 H])([2 H])N
IUPAC navn dideuterio-(3,4,5-trideuteriofuran-2-yl)methanamine
Molekylvægt (g/mol) 102.146
Formel vægt 102.0841 g/mol
Procent renhed 97 atom % D, min 98% Chemical Purity