Få mere at vide
Furfuryl-d5 Alcohol, TRC
Artikelnummer. 30408486
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30408486 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30408486 Leverandør Toronto Research Chemicals Leverandørnr. TRCF8642211MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Hydroxymethylfuran furfuryl-d5
CAS 1398065-62-7
Molekylær formel C5 2H5 H O2
Mængde 1 mg
Synonym 2-Furanmethan-α,α,3,4,5-d5-ol, Furfuryl alcohol-d5, 2-Hydroxymethylfuran-d5, 2-Hydroxymethylfuran-D5
InChI formel InChI=1S/C5H6O2/c6-4-5-2-1-3-7-5/h1-3,6H,4H2/i1D,2D,3D,4D2
SMIL [2H]c1oc(c([2H])c1[2H])C([2H])([2H])O
IUPAC navn dideuterio-(3,4,5-trideuteriofuran-2-yl)methanol
Molekylvægt (g/mol) 103.13
Formel vægt 103.0682