Få mere at vide
Furan-d4, TRC
Artikelnummer. 30406042
Skift visning
Mængde:
1 g
Artikelnummer. Mængde
30406042 1 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30406042 Leverandør Toronto Research Chemicals Leverandørnr. TRCF8636521G
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Furan D4
CAS 6142-90-1
Molekylær formel C4 2H4 O
Mængde 1 g
Synonym Furan-2,3,4,5-d4, Furan D4 (7CI,8CI,9CI), Perdeuteriofuran, Tetradeuteriofuran
InChI formel InChI=1S/C4H4O/c1-2-4-5-3-1/h1-4H/i1D,2D,3D,4D
SMIL [2H]c1oc([2H])c([2H])c1[2H]
IUPAC navn 2,3,4,5-tetradeuteriofuran
Molekylvægt (g/mol) 72.1
Formel vægt 72.0513