missing translation for 'onlineSavingsMsg'
Få mere at vide

Furan-2-ylmethyl-phenethyl-amine, TRC

Artikelnummer. 30402460
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30402460 50 mg
30400663 100 mg
30395955 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30402460 Leverandør Toronto Research Chemicals Leverandørnr. TRCF86420850MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 4439-55-8
Molekylær formel C13H15NO
Molekylvægt (g/mol) 201.26
Synonym N-(2-Phenylethyl)-2-furanmethanamine, (Furan-2-ylmethyl)(2-phenylethyl)amine, [(Furan-2-yl)methyl](2-phenylethyl)amine, N-phenethyl-Furfurylamine
SMIL C(Cc1ccccc1)NCc2occc2

Tekniske data

Kemisk navn eller materiale Furan-2-ylmethyl-phenethyl-amine
Mængde 50 mg
InChI formel InChI=1S/C13H15NO/c1-2-5-12(6-3-1)8-9-14-11-13-7-4-10-15-13/h1-7,10,14H,8-9,11H2
Formel vægt 201.1154
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.