Få mere at vide
Furamidine Dihydrochloride, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Furamidine Dihydrochloride |
| CAS | 55368-40-6 |
| Molekylær formel | C18H18Cl2N4O |
| Mængde | 5 mg |
| Synonym | 4,4'-(2,5-Furandiyl)bisbenzenecarboximidamide Dihydrochloride, 2,5-Bis(4-amidinophenyl)furan Dihydrochloride, 2,5-Bis(4-guanylphenyl)furan Dihydrochloride, WR 199385 |
| InChI formel | InChI=1S/C18H16N4O/c19-17(20)13-5-1-11(2-6-13)15-9-10-16(23-15)12-3-7-14(8-4-12)18(21)22/h1-10H,(H3,19,20)(H3,21,22) |
| SMIL | C1=CC(=CC=C1C2=CC=C(O2)C3=CC=C(C=C3)C(=N)N)C(=N)N |
| Molekylvægt (g/mol) | 377.27 |