Få mere at vide
Furaltadone, TRC
Artikelnummer. 30343970
Skift visning
Mængde:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30343970 100 mg
30333005 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30343970 Leverandør Toronto Research Chemicals Leverandørnr. TRCB497335100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Furaltadone
CAS 139-91-3
Molekylær formel C13 H16 N4 O6
Mængde 100 mg
Synonym 2-Oxazolidinone, 5-(4-morpholinylmethyl)-3-[[(5-nitro-2-furanyl)methylene]amino]-, 2-Oxazolidinone, 5-(morpholinomethyl)-3-[(5-nitrofurfurylidene)amino]- (6CI,7CI,8CI), 3-(5-Nitro-2-furfurylideneamino)-5-(4-morpholinomethyl)-2-oxazolidone, 5-(4-Morpholinomethyl)-3-(5-nitro-2-furfurylideneamino)-2-oxazolidinone, 5-(N-Morpholinomethyl)-3-(5-nitrofurfurylideneamino)-2-oxazolidone, 5-Morpholinomethyl-3-(5-nitrofurfurylidenamino)-2-oxazolidone, 5-Morpholinomethyl-3-(5-nitrofurfurylideneamino)-2-oxazolidinone, 5-Morpholinomethyl-3-(5-nitrofurfurylideneamino)-2-oxazolidone, 5-Morpholinomethyl-3-(5-nitrofurfurylideneamino)oxazolidone, Altabactina, Altafur, Anauran, F 150, Fur-Novo, Furaltadon, Furaltadone, Furantoina, Furazolin, Furazoline, Furitale, Furlate, Furlidon, Furmethanol, Furmethonol, Furmetonol, Furmetox, Ibifur, Medifuran, N-(5-Nitro-2-furylidene)-3-amino-5-(N'-morpholinomethyl)-2-oxazolidone, NF 260, NSC 42388, Nitraldone, Nitrofurmethone, Nitrofurmeton, Otifuril, Sepsinol, Ultrafur, Unifur, Valsyn
InChI formel InChI=1S/C13H16N4O6/c18-13-16(14-7-10-1-2-12(22-10)17(19)20)9-11(23-13)8-15-3-5-21-6-4-15/h1-2,7,11H,3-6,8-9H2
SMIL [O-][N+](=O)c1oc(C=NN2CC(CN3CCOCC3)OC2=O)cc1
IUPAC navn 5-(morpholin-4-ylmethyl)-3-[(5-nitrofuran-2-yl)methylideneamino]-1,3-oxazolidin-2-one
Molekylvægt (g/mol) 324.29
Formel vægt 324.107