Få mere at vide
Furaltadone, LoGiCal
Artikelnummer. 30463282
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30463282 10 mg
1 options
Artikelnummer. 30463282 Leverandør LoGiCal Leverandørnr. LGCFOR1753.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Furaltadone
CAS 139-91-3
Analyt- eller komponentnavne Furaltadone
Forsendelsestilstand Room Temperature
Molekylær formel C13 H16 N4 O6
Certificeringer/compliance ISO 9001
Synonym 2-Oxazolidinone, 5-(4-morpholinylmethyl)-3-[[(5-nitro-2-furanyl)methylene]amino]-, 2-Oxazolidinone, 5-(morpholinomethyl)-3-[(5-nitrofurfurylidene)amino]- (6CI,7CI,8CI), 3-(5-Nitro-2-furfurylideneamino)-5-(4-morpholinomethyl)-2-oxazolidone, 5-(4-Morpholinomethyl)-3-(5-nitro-2-furfurylideneamino)-2-oxazolidinone, 5-(N-Morpholinomethyl)-3-(5-nitrofurfurylideneamino)-2-oxazolidone, 5-Morpholinomethyl-3-(5-nitrofurfurylidenamino)-2-oxazolidone, 5-Morpholinomethyl-3-(5-nitrofurfurylideneamino)-2-oxazolidinone, 5-Morpholinomethyl-3-(5-nitrofurfurylideneamino)-2-oxazolidone, 5-Morpholinomethyl-3-(5-nitrofurfurylideneamino)oxazolidone, Altabactina, Altafur, Anauran, F 150, Fur-Novo, Furaltadon, Furaltadone, Furantoina, Furazolin, Furazoline, Furitale, Furlate, Furlidon, Furmethanol, Furmethonol, Furmetonol, Furmetox, Ibifur, Medifuran, N-(5-Nitro-2-furylidene)-3-amino-5-(N'-morpholinomethyl)-2-oxazolidone, NF 260, NSC 42388, Nitraldone, Nitrofurmethone, Nitrofurmeton, Otifuril, Sepsinol, Ultrafur, Unifur, Valsyn
InChI formel InChI=1S/C13H16N4O6/c18-13-16(14-7-10-1-2-12(22-10)17(19)20)9-11(23-13)8-15-3-5-21-6-4-15/h1-2,7,11H,3-6,8-9H2
Løsningstype Neat
SMIL [O-][N+](=O)c1oc(C=NN2CC(CN3CCOCC3)OC2=O)cc1
IUPAC navn 5-(morpholin-4-ylmethyl)-3-[(5-nitrofuran-2-yl)methylideneamino]-1,3-oxazolidin-2-one
Industry Type Clinical Testing
Molekylvægt (g/mol) 324.29
Mængde 10 mg
Formel vægt 324.107 g/mol